missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Stearate, NF, BP, JP, 4-5%, Spectrum™ Chemical

Catalog No. p-7157795
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604113 500 g Poly Bottle
18604115 2.5 kg Poly Pail
18604114 6 x 500 g Poly Bottle
18604116 4 x 2.5 kg Poly Pail
18604117 12 kg Fiber Drum
18604118 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18604113 Supplier Spectrum Chemical Mfg Cor Supplier No. MA130500GM
Only null left

MA130, 557-04-0

Spectrum™ Chemical Magnesium Stearate, NF, BP, JP is used as an anti-adherent in the manufacture of of medical capsules, tablets, and powders. Spectrum Chemical manufactured NF, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.26
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
IUPAC Name magnesium(2+) dioctadecanoate
SMILES [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 4 to 5%
Grade Multi-Compendial
Loss on Drying 6%
Packaging Poly Bottle