missing translation for 'onlineSavingsMsg'
Learn More

Magnesium salicylate tetrahydrate, tech.

Catalog No. p-7050742
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3937922 100 g
AA3937936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3937922 Supplier Thermo Scientific Chemicals Supplier No. 03937922
Only null left

CAS: 18917-95-8 | C14H18MgO10 | 370.59 g/mol

It is used to treat mild to moderate muscular pain. It is also used to treat headaches, general back pain, and certain joint pains like arthritis. It is found in a variety of over-the-counter (OTC) medications as an anti-inflammatory, primarily for back-pain relief.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18917-95-8
Molecular Formula C14H18MgO10
Molecular Weight (g/mol) 370.59
MDL Number MFCD01766176,MFCD00045815
InChI Key NBQBEWAYWAMLJJ-UHFFFAOYSA-L
Synonym magnesium salicylate
PubChem CID 131674114
IUPAC Name magnesium;2-hydroxybenzoic acid;2-oxidobenzoate;hydrate
SMILES O.O.O.O.[Mg++].OC1=CC=CC=C1C([O-])=O.OC1=CC=CC=C1C([O-])=O

Specifications

Odor Odorless
Quantity 100 g
Sensitivity Air Sensitive
Merck Index 14,5672
Solubility Information Soluble in water and alcohol
Formula Weight 370.60 (298.60 Anhydrous)
Grade Technical
Physical Form Powder
Chemical Name or Material Magnesium salicylate tetrahydrate
EINECSNumber 242-669-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only