missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium phosphate pentahydrate, P{2}O{5} 33%
Description
Magnesium phosphate pentahydrate is used in the preparation of magnesium calcium phosphate biocement(MCPB) for bone regeneration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystals |
| Quantity | 1 kg |
| Assay Percent Range | P2O5 33% |
| Solubility Information | Insoluble in water. |
| Formula Weight | 352.93 (262.86 Anhydrous) |
| Appearance | White crystals |
| Density | g/cm3 at 20°C |
| Chemical Name or Material | Magnesium phosphate pentahydrate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 231-824-0 |
| RTECSNumber | TC6380000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only