accessibility menu, dialog, popup

UserName

Explore All Our Special Offers, All in One Place

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 10233-87-1

H2OH2OH2OH2OH2OMgOPOOO

CAS RN 10233-87-1

IUPAC Name: magnesium(2+) pentahydrate phosphate
Synonyms: magnesium phosphate tribasic
Molecular Weight (g/mol): 209.35
Molecular Formula: H10MgO9P
InChi Key: ZHDVAZVXDZNFPS-UHFFFAOYSA-K
SMILES: O.O.O.O.O.[Mg++].[O-]P([O-])([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.