missing translation for 'onlineSavingsMsg'
Learn More

Magnesium phosphate pentahydrate, P{2}O{5} 33%

Catalog No. AA18219A1
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA18219A1 1 kg
AA18219A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA18219A1 Supplier Thermo Scientific Chemicals Supplier No. 018219A1
Only null left

CAS: 10233-87-1 | H10Mg3O13P2 | 352.93 g/mol

Magnesium phosphate pentahydrate is used in the preparation of magnesium calcium phosphate biocement(MCPB) for bone regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10233-87-1
Molecular Formula H10Mg3O13P2
Molecular Weight (g/mol) 352.93
MDL Number MFCD00150252
InChI Key QZYXGKYJDBUYES-UHFFFAOYSA-H
Synonym magnesium phosphate pentahydrate, magnesium phosphate usp, phosphoric acid, magnesium salt 2:3 , pentahydrate, 3mg.2po4.5h2o, trimagnesium phosphate pentahydrate, magnesium phosphate 3:2 pentahydrate, magnesium phosphate, tribasic pentahydrate, magnesium phosphate pentahydrate, p2o, trimagnesium 2+ pentahydrate diphosphate
PubChem CID 23724945
IUPAC Name trimagnesium;diphosphate;pentahydrate
SMILES O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Mg+2].[Mg+2].[Mg+2]

Specifications

Color White
Physical Form Crystals
Quantity 1 kg
Assay Percent Range P2O5 33%
Solubility Information Insoluble in water.
Formula Weight 352.93 (262.86 Anhydrous)
Appearance White crystals
Density g/cm3 at 20°C
Chemical Name or Material Magnesium phosphate pentahydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-824-0
RTECSNumber TC6380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only