missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide substrate, 10x10x1mm, polished one side, 100 orientation

Catalog No. p-5548806
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38503KT 1 Ea.
AA38503KU 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38503KT Supplier Thermo Scientific Chemicals Supplier No. 038503KT
Only null left

CAS: 1309-48-4 | MgO | 40.30 g/mol

Magnesium oxide substrate is used widely in high temperature superconductor. It is also used as deposition of ferro electric thin film coatings on magnesium oxide substrates. It is also growing applications for MgO in the field of plasma display panel (PDP) technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Color White
Physical Form Plate
Quantity 1 Ea.
Solubility Information Soluble in acid,ammonia. Insoluble in alcohol.
Odor Odorless
Chemical Name or Material Magnesium Oxide Substrate, Polished One Side
EINECSNumber 215-171-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only