missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation

Catalog No. p-7050299
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36427KT 1 Ea.
AA36427KU 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36427KT Supplier Thermo Scientific Chemicals Supplier No. 036427KT
Only null left

CAS: 1309-48-4 | MgO | 40.30 g/mol

A useful laboratory reagent, and a good reference white color for colorimetry. The interaction of magnesium oxide substrates with nickel ferrite films has been investigated using a wide range of electron optical techniques. Considerable interdiffusion is observed, which is likely to alter the magnetic properties of these films. Epitaxial ferrite-spinel layer is formed on magnesium oxide substrate. Additionally, Magnesium oxide in nanoparticles dispersed in alcohol have been reported to inhibit hydrolytic and oxidative degradation of paper documents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Quantity 1 Ea.
Solubility Information Soluble in 5 M HCl (0.1 M) at 20°C,and dilute acids. Insoluble in water,and ethanol.
Chemical Name or Material Magnesium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation
EINECSNumber 215-171-9
RTECSNumber OM3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only