missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 100 Orientation, Thermo Scientific™

$195.95 - $195.95

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA41436KT
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
41436KT
1 Ea.
Each for $195.95
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1309-48-4
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium

Specifications

White
Plate
MgO
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
O=[Mg]
40.30
Odorless
1309-48-4
1 Ea.
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
Magnesium Oxide Substrate, Polished Two Sides
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents