Learn More
Magnesium oxide, Puratronic™, 99.995% (metals basis)
CAS: 1309-48-4 | MgO | 40.30 g/mol
Supplier: Thermo Scientific Chemicals 01080018
Description
Magnesium oxide is used as a refractive material, which is physically and chemically stable at higher temperature. Magnesium oxide is a raw material for producing portland cement. It is also used in agricultural, chemical construction, environment al and other industrial applications. It is also used in medicine, colorimetry, industrial cables, heating elements, ceramics, spin-tunneling devices, and several other materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3600°C | |
| 2852°C | |
| 1309-48-4 | |
| 50 g | |
| MgO | |
| magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
| CPLXHLVBOLITMK-UHFFFAOYSA-N | |
| oxomagnesium | |
| 14792 | |
| 1.736 | |
| 3.58 g/cm3 at 20°C |
| Magnesium oxide | |
| White | |
| Powder | |
| 99.998% (Metal basis) | |
| MFCD00011109 | |
| Soluble in acid and ammonia. Insoluble in alcohol. | |
| O=[Mg] | |
| 40.30 | |
| 40.3 | |
| White powder |
Chemical Identifiers
| 1309-48-4 | |
| 40.30 | |
| CPLXHLVBOLITMK-UHFFFAOYSA-N | |
| 14792 | |
| O=[Mg] |
| MgO | |
| MFCD00011109 | |
| magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
| oxomagnesium |
Safety and Handling
EINECSNumber : 215-171-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only