missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide, Powder, U.S.P. - F.C.C., J.T. Baker™

Catalog No. p-5390784
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2003270 500 g Poly Bottle
02003269 2 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2003270 Supplier Avantor Supplier No. 248001
Only null left

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 40.3
Grade FCC/USP
Packaging Poly Bottle
Specific Gravity 1L = 3.58kg
Chemical Name or Material Magnesium Oxide