missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide, Light Powder, USP, 96-100.5%, Spectrum™ Chemical

MA120, 1309-48-4, MgO

Supplier:  Spectrum Chemical Mfg Cor MA12025KG

 View more versions of this product

Catalog No. 18604083


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Magnesium Oxide, Light Powder, USP has many uses, but from a pharmaceutical perspective it is used primarily as a mineral supplement and a tablet and capsule diluent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

1309-48-4
2.5 kg
MFCD00011109
O=[Mg]
40.30
USP
Poly Pail
100%
MgO
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
96 to 100.5%
10%

Chemical Identifiers

1309-48-4
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
O=[Mg]
MgO
MFCD00011109
oxomagnesium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions