missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide, Light Powder, USP, 96-100.5%, Spectrum™ Chemical

Catalog No. 18604082
Change view
Click to view available options
Quantity:
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604082 500 g Poly Pail
18604083 2.5 kg Poly Pail
18604085 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604082 Supplier Spectrum Chemical Mfg Cor Supplier No. MA120500GM

MA120, 1309-48-4, MgO

Spectrum™ Chemical Magnesium Oxide, Light Powder, USP has many uses, but from a pharmaceutical perspective it is used primarily as a mineral supplement and a tablet and capsule diluent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 96 to 100.5%
Grade USP
Loss on Ignition 10%
Packaging Poly Pail