missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide (Heavy Powder/USP/FCC), Fisher Chemical™

Catalog No. p-32366
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M68500 500 g Poly Bottle
M683 3 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M68500 Supplier Thermo Fisher Scientific Supplier No. M68 500
Only null left

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2800°C
Color White
pH 10.3
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 40.3
Percent Purity 96.0 to 100.5% (USP)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (FCC/USP), Pass Test (USP)
Loss on Ignition 10.0% max. (FCC), 10.0% max. (FCC/USP), 10.0% max. (USP)
Packaging Poly Bottle
Vapor Pressure 0 hPa at 20°C
Density 3.580g/cm³
Chemical Name or Material Magnesium Oxide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin and respiratory tract irritation.
NFPA
Health: 1
Flammability: 0
Instability: 1