missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Oxide, Heavy Powder, USP, 96-100.5%, Spectrum™ Chemical
MA125, 1309-48-4, MgO
$653.80 - $8169.40
Chemical Identifiers
| CAS | 1309-48-4 |
|---|---|
| Molecular Formula | MgO |
| Molecular Weight (g/mol) | 40.30 |
| MDL Number | MFCD00011109 |
| InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
| IUPAC Name | oxomagnesium |
| SMILES | O=[Mg] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604078
|
Spectrum Chemical Mfg Cor
MA125500GM |
500 g | Poly Bottle |
Each for $653.80
|
|
||||
|
18604079
|
Spectrum Chemical Mfg Cor
MA12525KG |
2.5 kg | Poly Pail |
Each for $1,818.30
|
|
||||
|
18604080
|
Spectrum Chemical Mfg Cor
MA12512KG |
12 kg | Poly Pail |
Each for $2,888.07
|
|
||||
|
18604081
|
Spectrum Chemical Mfg Cor
MA12525KGBL |
25 kg | Fiber Drum |
Each for $8,169.40
|
|
||||