missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 99.99%, (trace metal basis), extra pure

Catalog No. p-3776945
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC193430100 10 g Glass bottle
AC193430500 50 g Glass bottle
2 options

Catalog No. AC193430100

Supplier: Thermo Scientific Chemicals 193430100

Only null left

CAS: 1309-48-4 | MgO | 40.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600.0°C
Melting Point 2800.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99%
Linear Formula MgO
Fieser 01,633; 02,256; 07,221; 16,200
Merck Index 15, 5741
Solubility Information Solubility in water: 6.2 mg/l (20°C), reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids, insoluble in alcohol
Formula Weight 40.29
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 3.58
Density 3.5800g/mL
Chemical Name or Material Magnesium oxide, Extra Pure
Show More Show Less
Hazard Category
EINECSNumber 215-171-9

RUO – Research Use Only