missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 99.95% (metals basis)

Catalog No. AA4164814
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA4164814 25 g Glass bottle
AA4164822 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4164814 Supplier Thermo Scientific Chemicals Supplier No. 04164814

Used as a laboratory reagent, and a reference white color for colorimetry. Magneisum oxide microspheres is used for enrichment method of polycyclic aromatic hydrocarbons. It is also used in groundwater studies. | CAS: 1309-48-4 | MgO | 40.30 g/mol

Magnesium oxide is used as a laboratory reagent, and a good reference white color for colorimetry. Magneisum oxide microspheres is used for enrichment method of polycyclic aromatic hydrocarbons. It is also used in groundwater studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2852°C
Physical Form Fused Lump
Quantity 25 g
Solubility Information Soluble in HCl and dilute acids. Insoluble in water and ethanol.
Formula Weight 40.3
Percent Purity 99.95%
Packaging Glass bottle
Refractive Index 1.736
Specific Gravity 3.58
Density 3.58
Chemical Name or Material Magnesium oxide
Show More Show Less
EINECSNumber 215-171-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only