missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Boiling Point | 3600.0°C |
| Melting Point | 2800.0°C |
| Color | White |
| Physical Form | Amorphous Powder |
| Quantity | 500 g |
| Loss on Ignition | 2% max. |
| Packaging | Plastic bottle |
| Specific Gravity | 3.58 |
| Assay Percent Range | 97.5% min. (Complexometry) |
| Density | 3.5800g/mL |
| Show More |
Safety and Handling
| Hazard Category | |
| EINECSNumber | 215-171-9 |
RUO – Research Use Only