missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 97+%, for analysis

Catalog No. p-3212839
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Catalog No. Quantity Packaging
AC222532500 250 g Glass bottle
AC222530010 1 kg Plastic bottle
2 options

Catalog No. AC222532500

Supplier: Thermo Scientific Chemicals 222532500

Only null left

CAS: 1309-48-4 | MgO | 40.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2800°C
Color White
Physical Form Amorphous Powder
Quantity 250 g
Assay Percent Range 97% min. (on dry substance) (Complexometry)
Linear Formula MgO
Fieser 01,633; 02,256; 07,221; 16,200
Merck Index 15, 5741
Solubility Information Solubility in water: 6.2 mg/l (20°C), reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids, insoluble in alcohol
Formula Weight 40.29
Percent Purity ≥97%
Grade Analytical
Loss on Ignition 3.0% max.
Packaging Glass bottle
Specific Gravity 3.58
Density 3.5800g/mL
Chemical Name or Material Magnesium oxide
Show More Show Less
Hazard Category
EINECSNumber 215-171-9

RUO – Research Use Only