missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxalate dihydrate, 98.5+%

Catalog No. p-7059106
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4397622 100 g
AA4397636 500 g
AA43976A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4397622 Supplier Thermo Scientific Chemicals Supplier No. 04397622
Only null left

CAS: 547-66-0 | C2H2MgO4+2 | 114.339 g/mol

Magnesium oxalate dihydrate is in the synthesis of nano sized magnesium oxide. It is also used as a catalyst, refractory materials, adsorbents, superconductors, and ferroelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 547-66-0
Molecular Formula C2H2MgO4+2
Molecular Weight (g/mol) 114.339
MDL Number MFCD00287279
InChI Key UHNWOJJPXCYKCG-UHFFFAOYSA-N
Synonym magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1
PubChem CID 54611841
IUPAC Name magnesium;oxalic acid
SMILES C(=O)(C(=O)O)O.[Mg+2]

Specifications

Melting Point 150°C (decomposition)
Physical Form Powder, Pieces
Quantity 100 g
UN Number UN3288
Merck Index 14,5676
Solubility Information Soluble in diluted mineral acid,water (0.038g/100g H2O ). Insoluble in organics solutions.
Formula Weight 148.36 (112.32 Anhydrous)
Percent Purity ≥98.5%
Odor Odorless
Chemical Name or Material Magnesium oxalate dihydrate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 208-932-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only