CAS RN 547-66-0
IUPAC Name:
magnesium(2+) oxalate
Synonyms:
magnesium oxalate
Molecular Weight (g/mol):
112.32
Molecular Formula:
C2MgO4
InChi Key:
UHNWOJJPXCYKCG-UHFFFAOYSA-L
SMILES:
[Mg++].[O-]C(=O)C([O-])=O
1
–
1
of
1
results
1
Magnesium oxalate dihydrate, 98.5+%
CAS: 547-66-0 Molecular Formula: C2H2MgO4+2 Molecular Weight (g/mol): 114.339 MDL Number: MFCD00287279 InChI Key: UHNWOJJPXCYKCG-UHFFFAOYSA-N Synonym: magnesium oxalate,magnesium oxalate 1:1,magnesium oxalate mgc2o4,oxalic acid, magnesium salt 1:1,ethanedioic acid, magnesium salt 1:1 PubChem CID: 54611841 IUPAC Name: magnesium;oxalic acid SMILES: C(=O)(C(=O)O)O.[Mg+2]