missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1079906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1079906 5 g
AA1079914 25 g
AA1079922 100 g
AA1079936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1079906 Supplier Thermo Scientific Chemicals Supplier No. 01079906

CAS: 10377-60-3 | MgN2O6 | 148.31 g/mol

It is used in printing, chemical, agriculture and ceramics industries. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10377-60-3
Molecular Formula MgN2O6
Molecular Weight (g/mol) 148.31
MDL Number MFCD00011103
InChI Key YIXJRHPUWRPCBB-UHFFFAOYSA-N
Synonym magnesium nitrate, magnesium dinitrate, nitric acid, magnesium salt, unii-77cbg3un78, magnesium nitrate solution, nitric acid, magnesium salt 2:1, magniosan, hsdb 660, magnesium ii nitrate 1:2, magnesium standard for ic
PubChem CID 25212
ChEBI CHEBI:64736
IUPAC Name magnesium;dinitrate
SMILES [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330°C (decomposition)
Melting Point 89°C
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1474
Merck Index 14,5674
Solubility Information Freely soluble in water. Moderately soluble in ethanol and ammonia.
Formula Weight 148.32 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 1.46 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: MAGNESIUM NITRATE
EINECSNumber 233-826-7
RTECSNumber OM3756000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only