Learn More
Magnesium nitrate hexahydrate, ACS, 98.0-102.0%
As fertilizer, in manufacture of ammonium nitrate | CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol
Supplier: Thermo Scientific Chemicals 01156436
Description
It is used in printing, chemical, agriculture and ceramics industries. It is also used in the detection of selenium in plasma samples, and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 330°C (decomposition) | |
| 89°C | |
| 13446-18-9 | |
| 500 g | |
| H12MgN2O12 | |
| magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty | |
| MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
| magnesium;dinitrate;hexahydrate | |
| 202877 | |
| ACS | |
| 1.46 |
| Magnesium nitrate | |
| hexahydrate | |
| Crystalline | |
| 98.0-102.0% | |
| MFCD00149779 | |
| Very soluble in water,alcohol and liquid ammonia. | |
| O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 256.40 | |
| 256.41 (148.32 Anhydrous) | |
| Hygroscopic |
Chemical Identifiers
| 13446-18-9 | |
| 256.40 | |
| MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
| 202877 | |
| O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| H12MgN2O12 | |
| MFCD00149779 | |
| magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty | |
| magnesium;dinitrate;hexahydrate |
Safety and Handling
EINECSNumber : 233-826-7
RTECSNumber : OM3756000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only