missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hexahydrate, ACS, 98.0-102.0%

Catalog No. p-3257032
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1156418 50 g
AA1156436 500 g
AA11564A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1156418 Supplier Thermo Scientific Chemicals Supplier No. 01156418
Only null left

As fertilizer, in manufacture of ammonium nitrate | CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol

It is used in printing, chemical, agriculture and ceramics industries. It is also used in the detection of selenium in plasma samples, and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330°C (decomposition)
Melting Point 89°C
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 98.0-102.0%
Solubility Information Very soluble in water,alcohol and liquid ammonia.
Formula Weight 256.41 (148.32 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Density 1.46
Chemical Name or Material Magnesium nitrate, hexahydrate
Show More Show Less
EINECSNumber 233-826-7
RTECSNumber OM3756000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only