missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Matrix, Modifier for Graphite Furnace-AAS, MilliporeSigma™ Supelco™

Catalog No. 111018795
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018795 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018795 Supplier Merck Emd Millipore Supplier No. 6304350MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Matrix modification is a procedure for reducing volatilization and vapour-phase intrusions using a chemical reagent which acts as buffer enabling the use of higher pre-treated and atomization temperatures. The reagent causes either the interfering component to become more volatile or the analyte is converted to less volatile form. Magnesium is a first class modifier, forming oxides in the graphite furnace during heat pre-treatment. Magnesium added as nitrate has high thermal stability.

Specifications

Packaging Glass Bottle
CAS 13446-18-9
Concentration 10.0 g/L ±0.3 g/L Mg (in ˜17% nitric acid)
MDL Number MFCD00085308
Synonym Magnesium nitrate - nitric acid solution; Magnesium modifier
Quantity 50 mL
Grade AAS