missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Hydroxide, Purified, Reagents
Magnesium Hydroxide, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2238700500A5
Specifications
| 673°C | |
| Colorless/White | |
| Solid | |
| H2MgO2 | |
| Milk of Magnesia, Magnesium Dihydroxide | |
| [OH-].[OH-].[Mg++] | |
| 58.32 | |
| Poly Bottle |
| Magnesium Hydroxide, Purified | |
| 1309-42-8 | |
| 500 g | |
| MFCD00011104 | |
| VTHJTEIRLNZDEV-UHFFFAOYSA-L | |
| magnesium(2+) dihydroxide | |
| Pure | |
| 2.36 |
Chemical Identifiers
| 1309-42-8 | |
| 58.32 | |
| VTHJTEIRLNZDEV-UHFFFAOYSA-L | |
| magnesium(2+) dihydroxide |
| H2MgO2 | |
| MFCD00011104 | |
| Milk of Magnesia, Magnesium Dihydroxide | |
| [OH-].[OH-].[Mg++] |