missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Hydroxide, Purified, Reagents

Catalog No. p-200021079
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337188 2.5 kg
01337187 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337188 Supplier Labchem Inc Supplier No. C22387002.5KG
Only null left

Magnesium Hydroxide, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym Milk of Magnesia, Magnesium Dihydroxide
IUPAC Name magnesium(2+) dihydroxide
SMILES [OH-].[OH-].[Mg++]

Specifications

Boiling Point 673°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 2.36
Chemical Name or Material Magnesium Hydroxide, Purified