missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium gluconate hydrate, Thermo Scientific™
Chemical Identifiers
| CAS | 3632-91-5 |
|---|---|
| Molecular Formula | C6H11MgO7 |
| Molecular Weight (g/mol) | 219.45 |
| MDL Number | MFCD00150971 |
| InChI Key | GUOKWHRDVGUKOZ-UHFFFAOYNA-M |
| IUPAC Name | magnesium(2+) 2,3,4,5,6-pentahydroxyhexanoate |
| SMILES | [Mg++].OCC(O)C(O)C(O)C(O)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC464830250
|
N/A | 25 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC464831000
|
N/A | 100 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 3632-91-5 | |
| 219.45 | |
| GUOKWHRDVGUKOZ-UHFFFAOYNA-M | |
| [Mg++].OCC(O)C(O)C(O)C(O)C([O-])=O |
| C6H11MgO7 | |
| MFCD00150971 | |
| magnesium(2+) 2,3,4,5,6-pentahydroxyhexanoate |
Specifications
| 3632-91-5 | |
| MFCD00150971 | |
| GUOKWHRDVGUKOZ-UHFFFAOYNA-M | |
| magnesium(2+) 2,3,4,5,6-pentahydroxyhexanoate | |
| 414.61 |
| C6H11MgO7 | |
| Solubility aq. soln.: 9.6 g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
| [Mg++].OCC(O)C(O)C(O)C(O)C([O-])=O | |
| 219.45 | |
| Magnesium gluconate hydrate |
Safety and Handling
EINECSNumber : 222-848-2
RUO – Research Use Only