missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium gluconate hydrate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 464831000
| Quantity | 100 g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 3632-91-5 | |
| 219.45 | |
| GUOKWHRDVGUKOZ-UHFFFAOYNA-M | |
| [Mg++].OCC(O)C(O)C(O)C(O)C([O-])=O |
| C6H11MgO7 | |
| MFCD00150971 | |
| magnesium(2+) 2,3,4,5,6-pentahydroxyhexanoate |
Specifications
| Magnesium gluconate hydrate | |
| 100 g | |
| MFCD00150971 | |
| GUOKWHRDVGUKOZ-UHFFFAOYNA-M | |
| magnesium(2+) 2,3,4,5,6-pentahydroxyhexanoate | |
| 414.61 |
| 3632-91-5 | |
| C6H11MgO7 | |
| Solubility aq. soln.: 9.6 g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether | |
| [Mg++].OCC(O)C(O)C(O)C(O)C([O-])=O | |
| 219.45 |
Safety and Handling
EINECSNumber : 222-848-2