missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Fluoride, Purified, Approx. 85%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M1045500GM

 View more versions of this product

Catalog No. 18611425


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7783-40-6
500 g
Purified
1
∼85%
Poly Bottle

Chemical Identifiers

7783-40-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.