missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Fluoride, Purified, Approx. 85%, Spectrum™ Chemical

Catalog No. p-7165818
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18611423 125 g Poly Bottle
18611425 500 g Poly Bottle
18611426 2.5 kg Poly Bottle
18611424 12 kg Poly Pail
4 options

Catalog No. 18611423

Supplier: Spectrum Chemical Mfg Cor M1045125GM

Only null left

Chemical Identifiers

CAS 7783-40-6

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity ∼85%
Grade Purified
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.