missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Citrate, Anhydrous, USP, 14.5-16.4%, Spectrum™ Chemical
M2036, 3344-18-1, C12H10Mg3O14
Supplier: Spectrum Chemical Mfg Cor M203625KG
Description
Spectrum™ Chemical Magnesium Citrate, Anhydrous, USP is often used as a saline laxative to clean stool from the intestines before surgery or certain bowel procedures. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 3344-18-1 | |
| 5.0-9.0 | |
| C12H16Mg3O14 | |
| [Mg].[Mg].[Mg].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O | |
| 457.16 | |
| USP | |
| Poly Pail |
| 100% | |
| 2.5 kg | |
| AOHMFUYIHARAGR-UHFFFAOYSA-N | |
| bis(2-hydroxypropane-1,2,3-tricarboxylic acid) trimagnesium | |
| 14.5 to 16.4% | |
| 2% |
Chemical Identifiers
| 3344-18-1 | |
| 457.16 | |
| bis(2-hydroxypropane-1,2,3-tricarboxylic acid) trimagnesium |
| C12H16Mg3O14 | |
| AOHMFUYIHARAGR-UHFFFAOYSA-N | |
| [Mg].[Mg].[Mg].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O |