Learn More
Magnesium chloride, ultra dry, 99.9% (metals basis)
Widely used as a source of magnesium ion, co-factor for many enzymes, for dust control and road stabilization. Also used in wastewater treatment, wallboard, artificial seawater, feed supplement, textiles, paper, fireproofing agents, cements and refrigeration brine. | CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol
$480.16 - $480.16
Chemical Identifiers
| CAS | 7786-30-3 |
|---|---|
| Molecular Formula | Cl2Mg |
| Molecular Weight (g/mol) | 95.21 |
| MDL Number | MFCD00011106 |
| InChI Key | TWRXJAOTZQYOKJ-UHFFFAOYSA-L |
| Synonym | magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride |
| PubChem CID | 5360315 |
| ChEBI | CHEBI:6636 |
| IUPAC Name | magnesium;dichloride |
| SMILES | [Mg++].[Cl-].[Cl-] |
Description
It is widely used as a source of magnesium ion, co-factor for many enzymes, for dust control and road stabilization. Also used in wastewater treatment, wallboard, artificial seawater, feed supplement, textiles, paper, fireproofing agents, cements and refrigeration brine. It is an important coagulant, an elution buffer for antibody affinity column purifications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7786-30-3 | |
| 95.21 | |
| TWRXJAOTZQYOKJ-UHFFFAOYSA-L | |
| 5360315 | |
| magnesium;dichloride |
| Cl2Mg | |
| MFCD00011106 | |
| magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride | |
| CHEBI:6636 | |
| [Mg++].[Cl-].[Cl-] |
Specifications
| 714°C | |
| Beads | |
| 99.9% (metals basis) | |
| MFCD00011106 | |
| Soluble in water,methanol,ethanol and alcohol. | |
| [Mg++].[Cl-].[Cl-] | |
| 95.21 | |
| CHEBI:6636 | |
| Hygroscopic | |
| Magnesium chloride, ultra dry |
| 7786-30-3 | |
| 50 g | |
| Cl2Mg | |
| magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride | |
| TWRXJAOTZQYOKJ-UHFFFAOYSA-L | |
| magnesium;dichloride | |
| 5360315 | |
| 95.22 | |
| 2.32 g/mL |
Safety and Handling
EINECSNumber : 232-094-6
RTECSNumber : OM2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only