missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride, Hexahydrate, Crystal, EP, BP, USP, 98-101%, Spectrum™ Chemical
MA110, 7791-18-6, MgCl2·6H2O
$565.73 - $7728.83
Chemical Identifiers
| CAS | 7791-18-6 |
|---|---|
| Molecular Formula | Cl2H12MgO6 |
| Molecular Weight (g/mol) | 203.30 |
| MDL Number | MFCD00149781 |
| InChI Key | DHRRIBDTHFBPNG-UHFFFAOYSA-L |
| IUPAC Name | magnesium(2+) hexahydrate dichloride |
| SMILES | O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604041
|
Spectrum Chemical Mfg Cor
MA110500GMPL |
500 g | Poly Bottle |
Each for $565.73
|
|
||||
|
18604043
|
Spectrum Chemical Mfg Cor
MA11025KGPL |
2.5 kg | Poly Pail |
Each for $1,737.00
|
|
||||
|
18604042
|
Spectrum Chemical Mfg Cor
MA110500GMPLCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,104.93
|
|
||||
|
18604044
|
Spectrum Chemical Mfg Cor
MA11025KGPLCS4 |
4 x 2.5 kg | Poly Pail |
Case of 4 Each for $5,998.77
|
|
||||
|
18604045
|
Spectrum Chemical Mfg Cor
MA11012KG |
12 kg | Poly Pail |
Each for $5,185.70
|
|
||||
|
18604046
|
Spectrum Chemical Mfg Cor
MA11050KGBL |
50 kg | Fiber Drum |
Each for $7,728.83
|
|
||||
Description
Spectrum™ Chemical Magnesium Chloride, Hexahydrate, Crystal, USP, EP, BP is used as a nutritional supplement. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7791-18-6 | |
| 203.30 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| magnesium(2+) hexahydrate dichloride |
Specifications
| 7791-18-6 | |
| 4.5-7.0 | |
| Cl2H12MgO6 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| magnesium(2+) hexahydrate dichloride | |
| 98 to 101% | |
| Poly Bottle |
| 100% | |
| 500 g | |
| MFCD00149781 | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
| 203.30 | |
| Multi-Compendial |