missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride, Hexahydrate, Crystal, EP, BP, USP, 98-101%, Spectrum™ Chemical
MA110, 7791-18-6, MgCl2·6H2O
Supplier: Spectrum Chemical Mfg Cor MA11012KG
Description
Spectrum™ Chemical Magnesium Chloride, Hexahydrate, Crystal, USP, EP, BP is used as a nutritional supplement. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 7791-18-6 | |
| 4.5-7.0 | |
| Cl2H12MgO6 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| magnesium(2+) hexahydrate dichloride | |
| 98 to 101% | |
| Poly Pail |
| 100% | |
| 12 kg | |
| MFCD00149781 | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
| 203.30 | |
| Multi-Compendial |
Chemical Identifiers
| 7791-18-6 | |
| 203.30 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| magnesium(2+) hexahydrate dichloride |