Learn More
Magnesium Chloride Hexahydrate (Crystalline, USP/FCC), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLM35212
Specifications
| Magnesium Chloride Hexahydrate | |
| White | |
| 5 to 6.5 | |
| 2.5 kg | |
| MFCD00149781 | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
| 203.30 | |
| 99.0 to 105.0% (FCC) | |
| Pass Test (FCC), Pass Test (USP), Pass Test (FCC/USP) | |
| >106°C |
| 117°C | |
| 7791-18-6 | |
| Powder/Solid | |
| Cl2H12MgO6 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| magnesium(2+) hexahydrate dichloride | |
| 203.3 | |
| FCC/USP | |
| Poly Bottle | |
| 1.56g/mL |
Chemical Identifiers
| 7791-18-6 | |
| 203.30 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| magnesium(2+) hexahydrate dichloride |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and respiratory system. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:1