missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate (Crystalline, USP/FCC), Fisher Chemical™

Catalog No. p-32326
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M35500 500 g Poly Bottle
M35212 2.5 kg Poly Bottle
M3512 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M35500 Supplier Thermo Fisher Scientific Supplier No. FLM35 500
Only null left

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C
Color White
pH 4.5 to 7.0
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 203.3
Percent Purity 98.0 to 101.0% (USP)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (USP), Pass Test (FCC/USP)
Packaging Poly Bottle
Decomposition Information >106°C
Density 1.56g/mL
Chemical Name or Material Magnesium Chloride Hexahydrate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and respiratory system. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1