missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous

Catalog No. p-4089760
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197060100 10 g Plastic bottle
AC197060500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197060100 Supplier Thermo Scientific Chemicals Supplier No. 197060100
Only null left

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
IUPAC Name magnesium(2+) dichloride
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Boiling Point 1412.0°C
Melting Point 708.0°C
Color White
Physical Form Beads
Quantity 10 g
Assay Percent Range 99.9%
Linear Formula MgCl2
Fieser 11,310
Merck Index 15, 5726
Solubility Information Solubility in water: 540g/L (20°C)
Formula Weight 95.21
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Magnesium chloride, <10 mesh, anhydrous
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-094-6

RUO – Research Use Only