missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium carbonate hydroxide pentahydrate, light, 98%
CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol
$49.07 - $531.36
Chemical Identifiers
| CAS | 56378-72-4 |
|---|---|
| Molecular Formula | C4H12Mg5O19 |
| Molecular Weight (g/mol) | 485.65 |
| MDL Number | MFCD00149784 |
| InChI Key | SXKPPTNZIUYFJR-UHFFFAOYSA-D |
| Synonym | acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5 |
| IUPAC Name | pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate |
| SMILES | O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1807030
|
Thermo Scientific Chemicals
A1807030 |
250 g |
Each for $49.07
|
|
|||||
|
AAA180700B
|
Thermo Scientific Chemicals
A180700B |
1000 g |
Each for $146.16
|
|
|||||
|
AAA180700I
|
Thermo Scientific Chemicals
A180700I |
5000 g |
Each for $531.36
|
|
|||||
Description
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMagnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.
Solubility
Soluble in water and acids. Insoluble in alcohol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 56378-72-4 | |
| 485.65 | |
| SXKPPTNZIUYFJR-UHFFFAOYSA-D | |
| pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate |
| C4H12Mg5O19 | |
| MFCD00149784 | |
| acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5 | |
| O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
Specifications
| 56378-72-4 | |
| C4H12Mg5O19 | |
| MFCD00149784 | |
| Soluble in water and acids. Insoluble in alcohol. | |
| O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O | |
| 485.65 | |
| 98% | |
| Magnesium carbonate hydroxide pentahydrate, light |
| 250 g | |
| 4MgCO3·Mg(OH)2·5H2O | |
| acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5 | |
| SXKPPTNZIUYFJR-UHFFFAOYSA-D | |
| pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate | |
| 485.65 (395.58 Anhydrous) | |
| 2.16 |
Safety and Handling
EINECSNumber : 235-192-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only