accessibility menu, dialog, popup

UserName

CAS RN 56378-72-4

IUPAC Name: pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
Synonyms: pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
Molecular Weight (g/mol): 485.65
Molecular Formula: C4H12Mg5O19
InChi Key: SXKPPTNZIUYFJR-UHFFFAOYSA-D
SMILES: O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)