missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate hydroxide pentahydrate, light, 98%

Catalog No. AAA1807030
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1807030 250 g
AAA180700B 1000 g
AAA180700I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1807030 Supplier Thermo Scientific Chemicals Supplier No. A1807030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol

Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water and acids. Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 56378-72-4
Molecular Formula C4H12Mg5O19
Molecular Weight (g/mol) 485.65
MDL Number MFCD00149784
InChI Key SXKPPTNZIUYFJR-UHFFFAOYSA-D
Synonym acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
IUPAC Name pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Quantity 250 g
Density 2.16
Linear Formula 4MgCO3·Mg(OH)2·5H2O
Solubility Information Soluble in water and acids. Insoluble in alcohol.
Formula Weight 485.65 (395.58 Anhydrous)
Percent Purity 98%
Chemical Name or Material Magnesium carbonate hydroxide pentahydrate, light
EINECSNumber 235-192-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only