missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium carbonate hydroxide pentahydrate, for biochemistry, 40.0-43.5% Magnesium oxide

CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol

$166.35 - $166.35

Chemical Identifiers

CAS 56378-72-4
Molecular Formula C4H12Mg5O19
Molecular Weight (g/mol) 485.65
MDL Number MFCD00149784
InChI Key SXKPPTNZIUYFJR-UHFFFAOYSA-D
Synonym acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
IUPAC Name pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC448045000
SDS View Documents
Thermo Scientific Chemicals
448045000
500 g
Each for $166.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56378-72-4
485.65
SXKPPTNZIUYFJR-UHFFFAOYSA-D
pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
C4H12Mg5O19
MFCD00149784
acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

350.0°C
500 g
Mg(OH)2·5H2O
acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
485.65
40.0 to 43.5% (Magnesium o)
Magnesium carbonate hydroxide pentahydrate
56378-72-4
C4H12Mg5O19
MFCD00149784
SXKPPTNZIUYFJR-UHFFFAOYSA-D
pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
485.69
Biochemical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only