missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Carbonate, Heavy, FCC, 40-43.5%, Spectrum™ Chemical
M1237, 39409-82-0, Approx. 4MgCO3·Mg(OH)2·5H2O
$441.60 - $4,684.00
Chemical Identifiers
| CAS | 39409-82-0 |
|---|---|
| Molecular Formula | C4H10Mg5O18 |
| Molecular Weight (g/mol) | 467.63 |
| MDL Number | MFCD00149783 |
| InChI Key | YMKSTRLFWJQKFP-UHFFFAOYSA-D |
| IUPAC Name | pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate |
| SMILES | O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604015
|
Spectrum Chemical Mfg Cor
M1237500GM |
500 g | Poly Pail |
N/A
|
|
||||
|
18604016
|
Spectrum Chemical Mfg Cor
M123725KG |
2.5 kg | Poly Pail |
N/A
|
|
||||
|
18604017
|
Spectrum Chemical Mfg Cor
M123712KG |
12 kg | Fiber Drum |
N/A
|
|
||||
|
18604018
|
Spectrum Chemical Mfg Cor
M123725KGBL |
25 kg | Fiber Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Magnesium Carbonate, Heavy, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 39409-82-0 | |
| 467.63 | |
| YMKSTRLFWJQKFP-UHFFFAOYSA-D | |
| O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
| C4H10Mg5O18 | |
| MFCD00149783 | |
| pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate |
Specifications
| 39409-82-0 | |
| 500 g | |
| MFCD00149783 | |
| O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O | |
| 467.63 | |
| FCC |
| 100% | |
| C4H10Mg5O18 | |
| YMKSTRLFWJQKFP-UHFFFAOYSA-D | |
| pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate | |
| 40 to 43.5% | |
| Poly Pail |