missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Carbonate, Heavy, FCC, 40-43.5%, Spectrum™ Chemical

Catalog No. 18604015
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604015 500 g Poly Pail
18604016 2.5 kg Poly Pail
18604017 12 kg Fiber Drum
18604018 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604015 Supplier Spectrum Chemical Mfg Cor Supplier No. M1237500GM

M1237, 39409-82-0, Approx. 4MgCO3·Mg(OH)2·5H2O

Spectrum™ Chemical Magnesium Carbonate, Heavy, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 40 to 43.5%
Grade FCC
Packaging Poly Pail