missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Acetate Tetrahydrate, ACS Reagent, Reagents
Magnesium Acetate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$578.82 - $1836.39
Chemical Identifiers
| CAS | 16674-78-5 |
|---|---|
| Molecular Formula | C4H14MgO8 |
| Molecular Weight (g/mol) | 214.45 |
| MDL Number | MFCD00149214 |
| InChI Key | XKPKPGCRSHFTKM-UHFFFAOYSA-L |
| Synonym | Magnesium diacetate tetrahydrate |
| IUPAC Name | magnesium(2+) diacetate tetrahydrate |
| SMILES | O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-917
|
Labchem Inc
C12376001KG |
1 kg |
Each for $1,836.39
|
|
|||||
|
01-337-916
|
Labchem Inc
C1237600250A6 |
250 g |
Each for $578.82
|
|
|||||
Chemical Identifiers
| 16674-78-5 | |
| 214.45 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| magnesium(2+) diacetate tetrahydrate |
| C4H14MgO8 | |
| MFCD00149214 | |
| Magnesium diacetate tetrahydrate | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
Specifications
| Colorless/White | |
| Solid | |
| C4H14MgO8 | |
| Magnesium diacetate tetrahydrate | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O | |
| 214.45 | |
| Poly Bottle | |
| Magnesium Acetate Tetrahydrate, ACS Reagent |
| 16674-78-5 | |
| 1 kg | |
| MFCD00149214 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| magnesium(2+) diacetate tetrahydrate | |
| ACS | |
| 1.45 |