missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate Tetrahydrate, ACS Reagent, Reagents

Catalog No. 01337916
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
250 g
Catalog No. Quantity
01337916 250 g
01337917 1 kg
2 options

Catalog No. 01337916

Supplier: Labchem Inc C1237600250A6

Only null left

Magnesium Acetate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym Magnesium diacetate tetrahydrate
IUPAC Name magnesium(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 250 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.45
Chemical Name or Material Magnesium Acetate Tetrahydrate, ACS Reagent