missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Acetate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$15070.89 - $27599.67
Chemical Identifiers
| CAS | 16674-78-5 |
|---|---|
| Molecular Formula | C4H14MgO8 |
| Molecular Weight (g/mol) | 214.45 |
| MDL Number | MFCD00149214 |
| InChI Key | XKPKPGCRSHFTKM-UHFFFAOYSA-L |
| Synonym | magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p |
| PubChem CID | 134717 |
| SMILES | O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02003258
|
Avantor
242401 |
500 g | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
02003369
|
Avantor J.T.Baker
242405 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 16674-78-5 | |
| 214.45 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| 134717 |
| C4H14MgO8 | |
| MFCD00149214 | |
| magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
Specifications
| 16674-78-5 | |
| 500 g | |
| MFCD00149214 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| magnesium;diacetate;tetrahydrate | |
| 134717 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 1.45kg |
| Crystals | |
| C4H14MgO8 | |
| magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O | |
| 214.45 | |
| 214.45 | |
| Poly Bottle | |
| Magnesium Acetate, 4-Hydrate |