missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003369
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003369 2.5 kg Poly Bottle
02003258 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003369 Supplier Avantor Supplier No. 242405

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystals
Quantity 2.5 kg
Packaging Poly Bottle
Specific Gravity 1L = 1.45kg
IUPAC Name magnesium(2+) diacetate tetrahydrate
Formula Weight 214.45
Percent Purity 98 to 102%
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Magnesium Acetate, 4-Hydrate