missing translation for 'onlineSavingsMsg'
Learn More

Innovatek MagnaAmp™ G. stearothermophilus Self-Contained Biological Indicator

Innovatek MagnaAmp™ G. stearothermophilus Self-Contained Biological Indicator for monitoring the steam sterilization of liquids

$753.50 - $1,197.58

Specifications

Color Yellow Indicator
For Use With (Application) Monitoring pressure steam sterilization of liquids in larger volumes
Model MagnaAmp™
Temperature (Metric) 57°C
Heating Time 48 hr. at 37 °C
View More Specs

Products 2
Catalog Number Mfr. No. Population Price Quantity & Availability  
Catalog Number Mfr. No. Population Price Quantity & Availability  
361022495
View Documents Promotion Details
Innovatek
4140MA/5
105
Pack of 50 Each for $753.50
Only null left
 
361042548
View Documents Promotion Details
Innovatek
4140MA/6
106
Pack of 50 Each for $1,197.58
Only null left
 
Description

Description

  • MagnaAmp™ is a self-contained biological indicator ampoule for monitoring pressure steam sterilization of liquids in larger volumes
  • Each sealed glass ampoule contains bacterial spores in nutrient medium
  • Viable spore growth from a failed cycle will turn the broth bright yellow within 48 hours of incubation at 57°C
  • Includes ten negative control vials, containing only nutrient media and no spores
  • Use with MagnaAmp™ 14-cavity incubator (now with calibrated digital thermometer)
  • Unique neck design allows the vial to be suspended mid-flask, ensuring that conditions for complete sterilization have reached even the center of a body of liquid
Specifications

Specifications

Yellow Indicator
MagnaAmp™
48 hr. at 37 °C
50 Tests/Pk.
Monitoring pressure steam sterilization of liquids in larger volumes
57°C
10 Negative Control Vials
14 Cavities
Videos
SDS
Documents

Documents