missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral RNA Isolation Kit

Catalog No. AM1939
Encompass
Change view
Click to view available options
Quantity:
50 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1939 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1939 Supplier Applied Biosystems™ Supplier No. LSAM1939

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MagMAX™ Viral RNA Isolation kits have been optimized for use with biological fluids and cell-free samples such as serum, plasma, swabs, and cell culture media.

RNA isolation from cell-free samples presents some unique challenges compared to RNA isolation from tissues or cultured cells. These include low viral titers, large volumes to be processed, and sample complexity. Also, the procedure must be compatible with real-time qRT-PCR. The MagMAX™ Viral RNA isolation kits address all of these issues, without the need for organic solvents or RNA precipitation, eliminating the problems often encountered with filter-based RNA isolation methods such as clogging, large elution volumes, and inconsistent yields.

  • More effective recovery of viral RNA and DNA from diverse sample types
  • Improved qRT-PCR results
  • Ideal when working with low viral concentrations
  • Improved yields and reproducibility

MagMAX™ Viral RNA Isolation Kit

The improved version of the MagMAX™ Viral RNA Isolation Kit provides better viral particle lysis and more effective recovery of both viral RNA and DNA from a variety of cell-free biological samples, including plasma, sera, and swabs. The results of these changes are improved yields and reproducibility. Enhanced genomic DNA removal results in better qRT-PCR results.

The MagMAX™ Viral RNA Isolation Kit can efficiently isolate viral RNA and DNA from samples as large as 400 μL. RNA and DNA recovery is typically greater than 75%, but may vary depending on sample type. The RNA recovered with the kit is of high quality and purity and is suitable for real-time RT-PCR.

Accessory Products

Lysis/Binding Solution Concentrate, Wash Solution 1 Concentrate, Wash Solution 2 Concentrate, and Carrier RNA are components of the MagMAX™ Viral RNA Isolation Kits that may also be purchased separately.

Additional Processing Plates and Lids: we recommend 96-well plates and lids from Phenix Research Products (#MPU-8117, #MPE-8019).

Benefits of Magnetic Bead-based Isolation

Magnetic beads offer many benefits for isolating RNA from cells and viral RNA from cell-free samples. Ambion’s MagMAX™technology binds RNA more efficiently than glass fiber filter methods resulting in higher RNA yields. The RNA yields are more consistent, both from experiment to experiment, and over a broad range of sample sizes. MagMAX™magnetic bead technology eliminates filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high efficiency binding, the bound RNA can be eluted in just 20 to 50 μL of nuclease free water. This makes the MagMAX™ magnetic bead technology extremely useful for concentrating RNA from large, dilute samples.

Specifications

Content And Storage Processing Plate & Lid, Binding Solution Concentrate, Wash Solution 1 Concentrate, Wash Solution 2 Concentrate, Elution Buffer are all stored at room temperature. RNA Binding Beads are stored at 4°C and the Lysis ⁄Binding Enhancer is stored at –20°C.
Format Tube
Isolation Technology Magnetic Bead
Purification Time 45 min.
Sample Type Cell-free Biological Fluids, Liquid Samples (e.g. Serum), Plasma, Serum, Viral Samples
Final Product Type Viral DNA⁄RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
No. of Reactions 50
Product Line MagMAX
Quantity 50 preps
Scale Mini
Starting Material Amount Up to 400 μl of sample
Target Viral DNA/RNA
Yield 10 ng
Show More Show Less
Is the the direct lysis approach to DNA/RNA analysis applicable for extraction of viral RNA?

Some customers have isolated RNA from viruses. Unfortunately, the protein coat could cause problems. Another option would be to use the MagMax Viral Isolation Kit.

Can I edit the manufactured KingFisher Flex protocols?

No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.

Can I use magnetic beads from other vendors with the KingFisher Flex instrument?

For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.

I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?

Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.

Can I scale up my reaction volume with the KingFisher Flex instrument?

Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.

Are all of the Applied Biosystems MagMAX kits validated for the KingFisher Flex Magnetic Particle Processor?

Yes, all MagMAX kits can be used with the KingFisher Flex Magnetic Particle Processor.


For Veterinary Use Only.