missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen Wash Solution

Catalog No. A42360
Encompass
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42360 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42360 Supplier Applied Biosystems™ Supplier No. A42360

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This wash solution is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.

This wash solution is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more wash solution than is provided in the kit.

Important: This wash solution may develop inert white or brown particulates that float in solution. This is not a cause for concern and does not negatively affect performance.

Specifications

Sample Type Serum, Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Capacity 1000 mL
Content And Storage Store at room temperature.

For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Purification Target Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids
Quantity 1000 mL
Shipping Condition Room Temperature
I am seeing some small particles floating in the MagMAX Viral/Pathogen Wash Solution. Does this affect the performance of the wash solution?

The MagMAX Viral/Pathogen Wash Solution may develop inert white or brown particulates that float in solution. This is not a cause for concern and does not negatively affect performance. This statement is also listed on the product page: https://www.thermofisher.com/order/catalog/product/A42360

For Research Use Only. Not for use in diagnostic procedures.